Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Dsc3
Entrez Gene ID
13507
Gene Name
desmocollin 3
Gene Aliases
5430426I24Rik
Chromosome Location
Chr.18: 19960930 - 20002413 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001291809.1 NP_001278738.1 2 - 3 320 107
NM_007882.3 NP_031908.3 2 - 3 320 107
GenBank mRNA BC144845.1 - 2 - 3 305 107
BY745760.1 - 3 - 4 366 107
Y11169.1 - 2 - 3 320 107

View Full Product Details