The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_013923.2 | NP_038951.1 | 7 - 8 | 1780 | 62 |
GenBank mRNA | AF120206.1 | - | - | 1673 | 62 |
AF120207.1 | - | 6 - 7 | 1673 | 62 | |
AK147768.1 | - | 7 - 8 | 1768 | 62 | |
BC040769.1 | - | 6 - 7 | 1555 | 62 | |
DV656092.1 | - | 2 - 3 | 264 | 62 | |
X71642.1 | - | 3 - 4 | 327 | 62 |