The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NR_038073.1 | 3 - 4 | 163 | 104 | |
GenBank mRNA | AK041908.1 | - | 3 - 4 | 164 | 104 |
AK080350.1 | - | 3 - 4 | 131 | 104 |