Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Ints4
Entrez Gene ID
101861
Gene Name
integrator complex subunit 4
Gene Aliases
2610034N24Rik, AA692684, AW493223
Chromosome Location
Chr.7: 97480950 - 97541398 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_027256.2 NP_081532.1 19 - 20 2439 153
GenBank mRNA AK011676.1 - 5 - 6 436 153
BC013710.1 - 19 - 20 2385 153
BC013813.1 - 7 - 8 693 153
BC021838.1 - 12 - 13 1442 153
BC031145.1 - 13 - 14 1518 153

View Full Product Details