Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Wdr55
Entrez Gene ID
67936
Gene Name
WD repeat domain 55
Gene Aliases
2410080P20Rik, C80692
Chromosome Location
Chr.18: 36759739 - 36763708 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_026464.2 NP_080740.2 1 - 2 242 87
GenBank mRNA AK017695.1 - 1 - 2 220 87
AK018469.1 - 1 - 2 223 87
AK075958.1 - 1 - 2 242 87
AK146093.1 - 1 - 2 261 87
BC064750.1 - 1 - 2 202 87
BC131932.1 - 1 - 2 236 87
BC131934.1 - 1 - 2 236 87

View Full Product Details