Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Wdr55
Entrez Gene ID
67936
Gene Name
WD repeat domain 55
Gene Aliases
2410080P20Rik, C80692
Chromosome Location
Chr.18: 36759739 - 36763708 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_026464.2 NP_080740.2 3 - 4 429 67
GenBank mRNA AK017695.1 - 3 - 4 407 67
AK018469.1 - 3 - 4 410 67
AK075958.1 - 3 - 4 429 67
AK146093.1 - 3 - 4 448 67
BC011484.1 - 1 - 2 95 67
BC064750.1 - 3 - 4 66
BC131932.1 - 3 - 4 423 67
BC131934.1 - 3 - 4 423 67

View Full Product Details