The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_028036.3 | NP_082312.3 | 7 - 8 | 912 | 71 |
GenBank mRNA | AK010487.1 | - | 7 - 8 | 837 | 71 |
AK046207.1 | - | 7 - 8 | 950 | 71 | |
BC005637.1 | - | 7 - 8 | 914 | 71 |