Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Magi2
Entrez Gene ID
50791
Gene Name
membrane associated guanylate kinase, WW and PDZ domain containing 2
Gene Aliases
AIP-1, Acvri1, Acvrinp1, Acvrip1, Magi-2, S-SCAM, mKIAA0705
Chromosome Location
Chr.5: 19223283 - 20704792 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001170745.1 NP_001164216.1 16 - 17 3239 64
NM_001170746.1 NP_001164217.1 18 - 19 3446 64
NM_015823.3 NP_056638.1 17 - 18 3281 64
GenBank mRNA AK039336.1 - 14 - 15 2266 64
AK122349.1 - 16 - 17 2741 64
AK147530.1 - 16 - 17 3240 64
BC059005.1 - 17 - 18 2938 64

View Full Product Details