The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001168292.1 | NP_001161764.1 | 22 - 23 | 2686 | 82 |
NM_001168293.1 | NP_001161765.1 | 20 - 21 | 2905 | 82 | |
NM_027900.4 | NP_082176.4 | 20 - 21 | 2866 | 82 | |
GenBank mRNA | AK004884.1 | - | 10 - 11 | 1446 | 82 |
AK043545.1 | - | 20 - 21 | 2867 | 82 | |
AK122416.1 | - | 21 - 22 | 2544 | 82 | |
AX775759.1 | - | 11 - 12 | 1563 | 82 | |
BC023826.1 | - | 6 - 7 | 789 | 82 | |
BC043083.1 | - | 18 - 19 | 2244 | 82 | |
BC053099.1 | - | 15 - 16 | 1949 | 82 | |
BC064442.1 | - | 15 - 16 | 1917 | 82 | |
BC082999.1 | - | 8 - 9 | 1307 | 82 | |
BC117927.2 | - | 20 - 21 | 2648 | 82 |