Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Mmp19
Entrez Gene ID
58223
Gene Name
matrix metallopeptidase 19
Gene Aliases
-
Chromosome Location
Chr.10: 128790910 - 128800824 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001164197.1 NP_001157669.1 4 - 5 489 71
NM_021412.2 NP_067387.1 4 - 5 624 71
GenBank mRNA AF153199.1 - 4 - 5 576 71
AF155221.1 - 4 - 5 604 71
AF162446.1 - 4 - 5 604 71
AK079924.1 - 4 - 5 490 71
AK080125.1 - 4 - 5 484 71
AK086808.1 - 3 - 4 364 71
AK136517.1 - 4 - 5 478 71
AK154863.1 - 4 - 5 618 71
BC112966.2 - 4 - 5 596 71

View Full Product Details