The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_009299.2 | NP_033325.2 | 1 - 2 | 126 | 159 |
GenBank mRNA | BC104304.1 | - | 1 - 2 | 124 | 159 |
BC104305.1 | - | 1 - 2 | 126 | 159 | |
CJ055470.1 | - | 1 - 2 | 126 | 159 | |
M94179.1 | - | 1 - 2 | 112 | 159 |