Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- Srpk2
- Entrez Gene ID
-
20817
- Gene Name
- serine/arginine-rich protein specific kinase 2
- Gene Aliases
-
AW226533, AW492537, AW547358, Wbp6, mSRPK2
- Chromosome Location
-
Chr.5: 23503356 - 23684641 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AK161384.1 |
- |
1 - 2 |
235 |
76 |
View Full Product Details