Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Gmds
Entrez Gene ID
218138
Gene Name
GDP-mannose 4, 6-dehydratase
Gene Aliases
BC031788, C87208
Chromosome Location
Chr.13: 31819583 - 32338725 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_146041.2 NP_666153.1 10 - 11 1113 71
GenBank mRNA BC031788.1 - 10 - 11 1113 71
BC093502.1 - 10 - 11 1137 71

View Full Product Details