Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Zfpm2
Entrez Gene ID
22762
Gene Name
zinc finger protein, multitype 2
Gene Aliases
B330005D23Rik, FOG-2, FOG2
Chromosome Location
Chr.15: 40655042 - 41104592 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_011766.5 NP_035896.1 3 - 4 801 111
GenBank mRNA AF107306.1 - 3 - 4 322 111
AF118845.1 - 3 - 4 571 111
AF125166.1 - 3 - 4 802 111
AK046521.1 - 3 - 4 809 111
BC059241.1 - 3 - 4 701 111
EF514219.1 - 3 - 4 1624 111

View Full Product Details