Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Prl8a6
Entrez Gene ID
19112
Gene Name
prolactin family 8, subfamily a, member 6
Gene Aliases
PLP-C1, PLP-Ca, PLP-calpha, Prlpc, Prlpc1
Chromosome Location
Chr.13: 27432681 - 27438688 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001271379.1 NP_001258308.1 3 - 4 347 122
GenBank mRNA AK005478.1 - 3 - 4 348 122

View Full Product Details