Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Foxp2
Entrez Gene ID
114142
Gene Name
forkhead box P2
Gene Aliases
2810043D05Rik, AI449000, CAG-16, D0Kist7
Chromosome Location
Chr.6: 14901226 - 15441977 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001286607.1 NP_001273536.1 14 - 15 2026 77
NM_053242.4 NP_444472.2 16 - 17 2426 77
NM_212435.1 NP_997600.1 14 - 15 2089 77
GenBank mRNA AF339106.1 - 13 - 14 1767 77
AK082361.1 - 16 - 17 2098 77
AY079003.1 - 16 - 17 2055 77
BC058960.1 - 14 - 15 2023 77
BC062926.1 - 14 - 15 2089 77

View Full Product Details