Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- Slc17a1
- Entrez Gene ID
-
20504
- Gene Name
- solute carrier family 17 (sodium phosphate), member 1
- Gene Aliases
-
NAPI-1, Napi1, Npt1
- Chromosome Location
-
Chr.13: 23867723 - 23895730 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AI663290.1 |
- |
3 - 4 |
159 |
71 |
View Full Product Details