The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001252530.1 | NP_001239459.1 | 1 - 2 | 334 | 82 |
GenBank mRNA | AK087682.1 | - | 1 - 2 | 328 | 82 |
AK132394.1 | - | 1 - 2 | 335 | 82 | |
AK163542.1 | - | 1 - 2 | 330 | 83 |