The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_026458.4 | NP_080734.3 | 14 - 15 | 2176 | 96 |
GenBank mRNA | AK029665.1 | - | 15 - 16 | 2288 | 96 |
AY243470.1 | - | 14 - 15 | 2176 | 96 |