The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_177213.3 | NP_796187.2 | 28 - 29 | 4803 | 85 |
GenBank mRNA | AY243471.1 | - | 28 - 29 | 4803 | 85 |
BC141350.1 | - | 28 - 29 | 4768 | 85 | |
BC141351.1 | - | 28 - 29 | 4768 | 85 |