The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001113373.2 | NP_001106844.2 | 2 - 3 | 758 | 72 |
GenBank mRNA | AB099695.1 | - | 2 - 3 | 385 | 72 |
AK139360.1 | - | 2 - 3 | 759 | 72 |