Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- Mbtps2
- Entrez Gene ID
-
270669
- Gene Name
- membrane-bound transcription factor peptidase, site 2
- Gene Aliases
-
9630032G22Rik, AI662535
- Chromosome Location
-
Chr.X: 157547822 - 157598715 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AK077220.1 |
- |
- |
1634 |
64 |
View Full Product Details