The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001146294.1 | NP_001139766.1 | 9 - 10 | 1488 | 118 |
GenBank mRNA | BE861427.1 | - | 2 - 3 | 105 | 118 |