Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- B4galnt1
- Entrez Gene ID
-
14421
- Gene Name
- beta-1,4-N-acetyl-galactosaminyl transferase 1
- Gene Aliases
-
4933429D13Rik, Gal-NAc-T, GalNAc-T, GalNAcT, Galgt1, Ggm-2, Ggm2
- Chromosome Location
-
Chr.10: 127165156 - 127172340 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NR_045166.1 |
|
5 - 6 |
898 |
61 |
GenBank mRNA |
BC019969.1 |
- |
5 - 6 |
829 |
61 |
View Full Product Details