The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_009980.4 | NP_034110.1 | 3 - 4 | 611 | 113 |
GenBank mRNA | AB033123.1 | - | 3 - 4 | 476 | 113 |
AF059735.1 | - | 1 - 2 | 62 | 113 | |
AK146713.1 | - | 3 - 4 | 496 | 113 | |
AK147871.1 | - | 3 - 4 | 494 | 113 | |
AK178844.1 | - | 3 - 4 | 114 |