The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_177152.5 | NP_796126.4 | 8 - 9 | 1346 | 115 |
GenBank mRNA | AK129481.1 | - | 3 - 4 | 330 | 115 |
AK137007.1 | - | 8 - 9 | 1333 | 115 | |
AY505341.1 | - | 8 - 9 | 1289 | 115 | |
BC065142.1 | - | 8 - 9 | 1312 | 115 |