Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Fpr-rs4
Entrez Gene ID
14291
Gene Name
formyl peptide receptor, related sequence 4
Gene Aliases
-
Chromosome Location
Chr.17: 18021733 - 18022704 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_008041.2 NP_032067.2 1 - 1 68 124

View Full Product Details