Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Scgb1c1
Entrez Gene ID
338417
Gene Name
secretoglobin, family 1C, member 1
Gene Aliases
Ryd5
Chromosome Location
Chr.7: 140845565 - 140846769 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001099742.1 NP_001093212.1 1 - 2 65 63
GenBank mRNA AA462242.1 - 1 - 2 41 63
CB173308.1 - 1 - 2 42 63

View Full Product Details