The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_139294.5 | NP_647455.3 | 7 - 8 | 977 | 73 |
GenBank mRNA | AJ276307.1 | - | 5 - 6 | 661 | 73 |
AK140431.1 | - | 7 - 8 | 978 | 73 |