Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Avil
Entrez Gene ID
11567
Gene Name
advillin
Gene Aliases
Advil, DOC6
Chromosome Location
Chr.10: 127000075 - 127020994 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_009635.3 NP_033765.2 19 - 20 2490 81
GenBank mRNA AF041448.1 - 19 - 20 2441 81
AF059486.1 - 19 - 20 2460 81
AK154851.1 - 19 - 20 2461 81
AK155900.1 - 19 - 20 2461 81
AK171319.1 - 19 - 20 2462 81
BC119222.1 - 19 - 20 2395 81
BC120545.1 - 19 - 20 2395 81

View Full Product Details