The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_032542.1 | NP_115931.1 | 2 - 3 | 194 | 163 |
GenBank mRNA | AB050614.1 | - | 2 - 3 | 194 | 163 |
BC119343.1 | - | 2 - 3 | 294 | 163 | |
BC119345.1 | - | 2 - 3 | 294 | 163 |