Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Acsm1
Entrez Gene ID
117147
Gene Name
acyl-CoA synthetase medium-chain family member 1
Gene Aliases
Acas3, Bucs1, Macs
Chromosome Location
Chr.7: 119617756 - 119662515 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_054094.5 NP_473435.1 4 - 5 689 84
GenBank mRNA AB059428.1 - 4 - 5 693 84
AK149550.1 - 4 - 5 699 84
AK149586.1 - 4 - 5 715 84
BC016414.1 - 4 - 5 686 84
BC022149.1 - 4 - 5 667 84

View Full Product Details