Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Acsm1
Entrez Gene ID
117147
Gene Name
acyl-CoA synthetase medium-chain family member 1
Gene Aliases
Acas3, Bucs1, Macs
Chromosome Location
Chr.7: 119617756 - 119662515 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_054094.5 NP_473435.1 9 - 10 1271 64
GenBank mRNA AB059428.1 - 9 - 10 1275 64
AK149550.1 - 9 - 10 1281 64
BC016414.1 - 9 - 10 1268 64
BC022149.1 - 9 - 10 1249 64

View Full Product Details