The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001037865.1 | NP_001032954.1 | 33 - 34 | 3187 | 70 |
GenBank mRNA | AJ890449.1 | - | 33 - 34 | 3187 | 70 |
BC145537.1 | - | 32 - 33 | 3101 | 70 |