The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001012311.2 | NP_001012311.2 | 2 - 3 | 616 | 65 |
NM_001162919.1 | NP_001156391.1 | 2 - 3 | 674 | 65 | |
NM_001162920.1 | NP_001156392.1 | 2 - 3 | 616 | 65 | |
GenBank mRNA | AK076559.1 | - | 2 - 3 | 583 | 65 |
AK132880.1 | - | 2 - 3 | 675 | 65 | |
BC059882.1 | - | 2 - 3 | 607 | 65 | |
BC089360.1 | - | 2 - 3 | 464 | 65 | |
BY706951.1 | - | 2 - 3 | 616 | 65 |