The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_021435.3 | NP_067410.1 | 9 - 10 | 947 | 65 |
GenBank mRNA | AB041549.1 | - | 9 - 10 | 948 | 65 |
AK029046.1 | - | 8 - 9 | 3568 | 65 | |
AK029969.1 | - | 9 - 10 | 1005 | 65 | |
AK033913.1 | - | 9 - 10 | 877 | 65 | |
AK042150.1 | - | 9 - 10 | 847 | 65 | |
AK045791.1 | - | 9 - 10 | 890 | 65 | |
AK138591.1 | - | 9 - 10 | 888 | 65 | |
AK163299.1 | - | 4 - 5 | 415 | 65 | |
BC016275.1 | - | 9 - 10 | 878 | 65 |