Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Ubqln3
Entrez Gene ID
244178
Gene Name
ubiquilin 3
Gene Aliases
4933400K24Rik
Chromosome Location
Chr.7: 104140621 - 104143321 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_198623.2 NP_941025.1 1 - 2 72 100
GenBank mRNA AK077072.1 - 1 - 2 74 100
BC100419.1 - 1 - 2 49 100

View Full Product Details