The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_172814.3 | NP_766402.1 | 1 - 2 | 155 | 96 |
GenBank mRNA | AK050512.1 | - | 1 - 2 | 156 | 96 |