Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Tmc7
Entrez Gene ID
209760
Gene Name
transmembrane channel-like gene family 7
Gene Aliases
1700030H01Rik, C230064B05, C630024K23Rik
Chromosome Location
Chr.7: 118535841 - 118584790 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_172476.4 NP_766064.2 12 - 13 1750 74
GenBank mRNA AK083189.1 - 12 - 13 1753 74
AK132820.1 - 12 - 13 1780 74
AY236499.1 - 12 - 13 1811 74
AY263161.1 - 12 - 13 1779 74
BC105966.1 - 6 - 7 781 74

View Full Product Details