Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Ttc26
Entrez Gene ID
264134
Gene Name
tetratricopeptide repeat domain 26
Gene Aliases
9330141E21Rik, 9430097H08Rik
Chromosome Location
Chr.6: 38381474 - 38427647 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_153600.2 NP_705828.2 8 - 9 860 126
GenBank mRNA AK035185.1 - 8 - 9 868 126
BC066060.1 - 8 - 9 860 126
BC066087.1 - 9 - 10 1050 126

View Full Product Details