The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001252382.1 | NP_001239311.1 | 5 - 6 | 687 | 147 |
NM_012057.4 | NP_036187.1 | 5 - 6 | 635 | 147 | |
GenBank mRNA | AF028725.1 | - | 5 - 6 | 527 | 147 |
AK151019.1 | - | 5 - 6 | 552 | 147 | |
AK156220.1 | - | 5 - 6 | 687 | 147 | |
AK171356.1 | - | 5 - 6 | 555 | 147 |