Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Phldb1
Entrez Gene ID
102693
Gene Name
pleckstrin homology-like domain, family B, member 1
Gene Aliases
AU041016, D330037A14Rik, Ll5a
Chromosome Location
Chr.9: 44686308 - 44736151 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_153537.4 NP_705765.3 9 - 10 2485 91
GenBank mRNA AK122336.1 - 6 - 7 2203 91
BC058712.1 - 9 - 10 2481 91

View Full Product Details