The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_007619.2 | NP_031645.2 | 15 - 16 | 2604 | 63 |
GenBank mRNA | AK153915.1 | - | 15 - 16 | 2556 | 63 |
AK163907.1 | - | 14 - 15 | 2431 | 63 | |
BC125285.1 | - | 15 - 16 | 2519 | 63 | |
X57111.1 | - | 15 - 16 | 2491 | 63 |