Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Mcu
Entrez Gene ID
215999
Gene Name
mitochondrial calcium uniporter
Gene Aliases
2010012O16Rik, AV064928, C10orf42, Ccdc109a, D130073L02Rik, Gm64
Chromosome Location
Chr.10: 59446984 - 59616692 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001033259.4 NP_001028431.2 4 - 5 516 92
GenBank mRNA AK144727.1 - 4 - 5 517 92

View Full Product Details