Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Oit3
Entrez Gene ID
18302
Gene Name
oncoprotein induced transcript 3
Gene Aliases
EF-9, LZP
Chromosome Location
Chr.10: 59422960 - 59441779 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_010959.2 NP_035089.1 8 - 9 1562 84
GenBank mRNA AF356506.1 - 8 - 9 1509 84
AK040570.1 - 8 - 9 1563 84
AK050081.1 - 8 - 9 1529 84
AK153440.1 - 4 - 5 804 84
AY180915.1 - 8 - 9 1486 84

View Full Product Details