The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_027394.2 | NP_081670.1 | 9 - 10 | 1188 | 66 |
GenBank mRNA | AK011446.1 | - | 9 - 10 | 1205 | 66 |
AK049229.1 | - | 9 - 10 | 1189 | 66 | |
BC115686.1 | - | 9 - 10 | 1157 | 66 | |
BC115687.1 | - | 9 - 10 | 1156 | 66 |