Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Pmm2
Entrez Gene ID
54128
Gene Name
phosphomannomutase 2
Gene Aliases
AI585868, C86848
Chromosome Location
Chr.16: 8637707 - 8657552 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_016881.2 NP_058577.1 1 - 2 120 92
GenBank mRNA AF043514.1 - 1 - 2 120 92
AK002858.1 - 1 - 2 92 92
AK003334.1 - 1 - 2 78 92
AK005074.1 - 1 - 2 102 92
AK134850.1 - - 72 92
AK145402.1 - 1 - 2 72 92
AK149116.1 - 1 - 2 82 92
AK162626.1 - 1 - 2 90 92
BC037101.1 - 1 - 2 97 92
BC046325.1 - 1 - 2 68 92
BC094448.1 - 1 - 2 86 92

View Full Product Details