Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Pmm2
Entrez Gene ID
54128
Gene Name
phosphomannomutase 2
Gene Aliases
AI585868, C86848
Chromosome Location
Chr.16: 8637707 - 8657552 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_016881.2 NP_058577.1 4 - 5 404 85
GenBank mRNA AF043514.1 - 4 - 5 404 85
AK002858.1 - 4 - 5 346 85
AK003334.1 - 4 - 5 362 85
AK005074.1 - 4 - 5 386 85
AK012683.1 - 4 - 5 388 85
AK134850.1 - - 356 85
AK145402.1 - 4 - 5 356 85
AK149116.1 - 4 - 5 366 85
BC037101.1 - 4 - 5 381 85
BC046325.1 - 4 - 5 352 85
BC094448.1 - 4 - 5 370 85

View Full Product Details