The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001145884.1 | NP_001139356.1 | 7 - 8 | 1331 | 70 |
NM_010580.2 | NP_034710.2 | 7 - 8 | 1331 | 70 | |
GenBank mRNA | AF022110.1 | - | 7 - 8 | 1330 | 70 |
AF043256.1 | - | 7 - 8 | 1236 | 70 | |
AF043257.1 | - | 7 - 8 | 1236 | 70 | |
AK087476.1 | - | 7 - 8 | 1077 | 70 | |
AK140251.1 | - | 7 - 8 | 1243 | 70 | |
AK143532.1 | - | 7 - 8 | 1266 | 70 | |
BC029170.1 | - | 2 - 3 | 177 | 70 | |
BC046280.1 | - | 7 - 8 | 1240 | 70 | |
BC058246.1 | - | 7 - 8 | 1295 | 70 |