The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_198599.2 | NP_941001.2 | 2 - 3 | 539 | 99 |
GenBank mRNA | AK163656.1 | - | 2 - 3 | 503 | 99 |
BC116220.1 | - | 2 - 3 | 502 | 99 | |
BC116221.1 | - | 2 - 3 | 502 | 99 | |
BE979311.1 | - | 2 - 3 | 213 | 99 |